Assay Details
Gene Symbol: Mastl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107369.1 | NP_001100839.1 | 2398 | 95 | ||
XM_006254340.2 | XP_006254402.1 | 2463 | 95 | |||
XM_006254341.2 | XP_006254403.1 | 2325 | 95 | |||
GenBank mRNA | FQ224658.1 | - | 2557 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
