Assay Details
Gene Symbol: Mastl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107369.1 | NP_001100839.1 | 2284 | 59 | ||
XM_006254340.2 | XP_006254402.1 | 2349 | 59 | |||
XM_006254341.2 | XP_006254403.1 | 2211 | 59 | |||
GenBank mRNA | FQ224658.1 | - | 2443 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
