Assay Details
Gene Symbol: Hepacam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106580.2 | NP_001100050.1 | 6 - 7 | 1196 | 74 | |
XM_006236029.2 | XP_006236091.1 | 6 - 7 | 1249 | 74 | ||
GenBank mRNA | BC168741.1 | - | 6 - 7 | 1196 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
