Assay Details
Gene Symbol: Akap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227848.2 | XP_006227910.1 | 1 - 2 | 628 | 96 | |
GenBank mRNA | AY695058.1 | - | 1 - 2 | 151 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY695058.1, XM_006227848.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
