Assay Details
Gene Symbol: Slc7a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134615.1 | NP_001128087.1 | 7 - 8 | 2292 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017505 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P087856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124901 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017505 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P087856 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017505 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P087856 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P087856 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017505 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10814657 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10814656 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10814658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10814660 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10814655 | - | - |
Rat Genome 230 2.0 Array | 1378904_at | - | - |
Rat Genome 230 2.0 Array | 1378904_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378904_at | - | - |
Affymetrix HT Rat Focus Array | 1378904_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200005617.rn.1 | TC0200000926.rn.1 | RTA TC |
Rat Expression Set 230 | 1378904_PM_at | - | - |
Rat Expression Set 230 | 1378904_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21339814 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350426 | - | - |
