Assay Details
Gene Symbol: Nceh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127524.2 | NP_001120996.1 | 2 - 3 | 415 | 79 | |
GenBank mRNA | BC166533.1 | - | 2 - 3 | 415 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
