Assay Details
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037183.1 | NP_001032260.1 | 2 - 3 | 256 | 150 | |
GenBank mRNA | BC089884.1 | - | 2 - 3 | 257 | 150 | |
BC105820.1 | - | 2 - 3 | 256 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
