Assay Details
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037183.1 | NP_001032260.1 | 3 - 4 | 316 | 95 | |
XM_008760912.1 | XP_008759134.1 | 2 - 3 | 153 | 95 | ||
GenBank mRNA | BC089884.1 | - | 3 - 4 | 317 | 95 | |
BC105820.1 | - | 3 - 4 | 316 | 95 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008760912.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
