Assay Details
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037183.1 | NP_001032260.1 | 4 - 5 | 416 | 73 | |
XM_008760912.1 | XP_008759134.1 | 3 - 4 | 253 | 73 | ||
GenBank mRNA | BC089884.1 | - | 4 - 5 | 417 | 73 | |
BC105820.1 | - | 4 - 5 | 416 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
