Assay Details
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109097.1 | NP_001102567.1 | 4 - 5 | 579 | 76 | |
XM_006251655.2 | XP_006251717.1 | 4 - 5 | 626 | 76 | ||
GenBank mRNA | FQ214748.1 | - | 4 - 5 | 579 | 76 | |
FQ214808.1 | - | 4 - 5 | 579 | 76 | ||
FQ214853.1 | - | 4 - 5 | 562 | 76 | ||
FQ215077.1 | - | 581 | 76 | |||
FQ215190.1 | - | 4 - 5 | 581 | 76 | ||
FQ215360.1 | - | 4 - 5 | 585 | 76 | ||
FQ215728.1 | - | 4 - 5 | 581 | 76 | ||
FQ215901.1 | - | 4 - 5 | 583 | 76 | ||
FQ215918.1 | - | 4 - 5 | 76 | |||
FQ216019.1 | - | 4 - 5 | 619 | 76 | ||
FQ216115.1 | - | 4 - 5 | 580 | 76 | ||
FQ216248.1 | - | 4 - 5 | 580 | 76 | ||
FQ216396.1 | - | 4 - 5 | 563 | 76 | ||
FQ216753.1 | - | 4 - 5 | 563 | 76 | ||
FQ216842.1 | - | 4 - 5 | 563 | 76 | ||
FQ217380.1 | - | 601 | 76 | |||
FQ224894.1 | - | 4 - 5 | 583 | 76 | ||
FQ224977.1 | - | 4 - 5 | 579 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
