Assay Details
Gene Symbol: Abhd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007680.1 | NP_001007681.1 | 7 - 8 | 806 | 99 | |
XM_006251766.2 | XP_006251828.1 | 7 - 8 | 2085 | 99 | ||
XM_006251767.2 | XP_006251829.1 | 7 - 8 | 1617 | 99 | ||
XM_006251768.2 | XP_006251830.1 | 7 - 8 | 782 | 99 | ||
GenBank mRNA | BC083826.1 | - | 7 - 8 | 806 | 99 | |
FQ219067.1 | - | 2 - 3 | 84 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
