Assay Details
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012195.1 | NP_001012195.1 | 1 - 2 | 107 | 90 | |
XM_006240772.2 | XP_006240834.1 | 3 - 4 | 478 | 90 | ||
XM_006240773.2 | XP_006240835.1 | 3 - 4 | 540 | 90 | ||
XM_006240774.2 | XP_006240836.1 | 2 - 3 | 386 | 90 | ||
XM_006240775.2 | XP_006240837.1 | 3 - 4 | 396 | 90 | ||
XM_006240776.2 | XP_006240838.1 | 2 - 3 | 304 | 90 | ||
XM_008765030.1 | XP_008763252.1 | 2 - 3 | 1397 | 90 | ||
GenBank mRNA | BC083614.1 | - | 1 - 2 | 107 | 90 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083614.1, NM_001012195.1, XM_006240772.2, XM_006240773.2, XM_006240774.2, XM_006240775.2, XM_006240776.2, XM_008765030.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
