Assay Details
Gene Symbol: Eri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014143.1 | NP_001014165.1 | 2 - 3 | 395 | 115 | |
GenBank mRNA | BC089828.1 | - | 2 - 3 | 395 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
