Assay Details
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108203.1 | NP_001101673.1 | 2 - 3 | 160 | 75 | |
XM_006244524.2 | XP_006244586.1 | 2 - 3 | 215 | 75 | ||
XM_006244525.2 | XP_006244587.1 | 2 - 3 | 215 | 75 | ||
XM_006244527.2 | XP_006244589.1 | 2 - 3 | 215 | 75 | ||
XM_006244528.2 | XP_006244590.1 | 2 - 3 | 215 | 75 | ||
XM_006244534.2 | XP_006244596.1 | 2 - 3 | 215 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108203.1, XM_006244524.2, XM_006244525.2, XM_006244527.2, XM_006244528.2, XM_006244534.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
