Assay Details
Gene Symbol: Pla2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009353.1 | NP_001009353.1 | 2 - 3 | 329 | 76 | |
XM_006244606.2 | XP_006244668.1 | 2 - 3 | 446 | 76 | ||
GenBank mRNA | BC088457.1 | - | 2 - 3 | 329 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
