Assay Details
Gene Symbol: Pla2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009353.1 | NP_001009353.1 | 3 - 4 | 452 | 71 | |
XM_006244606.2 | XP_006244668.1 | 3 - 4 | 569 | 71 | ||
XM_008766856.1 | XP_008765078.1 | 2 - 3 | 2212 | 71 | ||
GenBank mRNA | BC088457.1 | - | 3 - 4 | 452 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
