Assay Details
Gene Symbol: LOC100911260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229638.1 | XP_006229700.1 | 11 - 12 | 3536 | 76 |
Gene Symbol: Hcfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139507.1 | NP_001132979.1 | 22 - 23 | 6673 | 76 | |
XM_006229593.2 | XP_006229655.1 | 22 - 23 | 6787 | 76 | ||
XM_006229594.2 | XP_006229656.1 | 22 - 23 | 6783 | 76 | ||
XM_006229596.2 | XP_006229658.1 | 22 - 23 | 6652 | 76 | ||
XM_006229597.1 | XP_006229659.1 | 22 - 23 | 6668 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
