Assay Details
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107339.1 | NP_001100809.1 | 4 - 5 | 695 | 82 | |
XM_008771495.1 | XP_008769717.1 | 3 - 4 | 407 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
