Assay Details
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022851.1 | NP_074042.1 | 3 - 4 | 595 | 63 | |
GenBank mRNA | FM032642.1 | - | 3 - 4 | 62 | ||
L34821.1 | - | 3 - 4 | 474 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
