Assay Details
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022851.1 | NP_074042.1 | 4 - 5 | 703 | 82 | |
GenBank mRNA | L34821.1 | - | 4 - 5 | 582 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
