Assay Details
Gene Symbol: Gfi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107823.1 | NP_001101293.1 | 5 - 6 | 767 | 68 | |
XM_008761651.1 | XP_008759873.1 | 5 - 6 | 854 | 68 | ||
GenBank mRNA | BC158685.1 | - | 5 - 6 | 782 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
