Assay Details
Gene Symbol: Mvb12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271232.1 | NP_001258161.1 | 3 - 4 | 363 | 115 | |
XM_008761761.1 | XP_008759983.1 | 3 - 4 | 419 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
