Assay Details
Gene Symbol: Mvb12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271232.1 | NP_001258161.1 | 6 - 7 | 716 | 51 | |
XM_008761761.1 | XP_008759983.1 | 6 - 7 | 772 | 51 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
