Assay Details
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100533.1 | NP_001094003.1 | 2 - 3 | 1010 | 71 | |
XM_006233679.2 | XP_006233741.1 | 2 - 3 | 8109 | 71 | ||
XM_008761623.1 | XP_008759845.1 | 1 - 2 | 2213 | 71 | ||
GenBank mRNA | BC092608.1 | - | 1 - 2 | 814 | 71 | |
CO395460.1 | - | 1 - 2 | 556 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
