Assay Details
Gene Symbol: Nr5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021742.1 | NP_068510.1 | 3 - 4 | 354 | 80 | |
XM_006249928.2 | XP_006249990.1 | 2 - 3 | 462 | 80 | ||
GenBank mRNA | AB012960.1 | - | 3 - 4 | 354 | 80 | |
AB012961.1 | - | 2 - 3 | 291 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
