Assay Details
Gene Symbol: Nmnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048042.1 | NP_001041507.1 | 8 - 9 | 787 | 84 | |
XM_008769614.1 | XP_008767836.1 | 8 - 9 | 645 | 84 | ||
GenBank mRNA | DQ022370.1 | - | 8 - 9 | 787 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
