Assay Details
Gene Symbol: Arhgef37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100974.1 | NP_001094444.1 | 10 - 11 | 1594 | 103 | |
XM_006254807.2 | XP_006254869.1 | 10 - 11 | 1653 | 103 | ||
XM_006254808.2 | XP_006254870.1 | 10 - 11 | 3593 | 103 | ||
XM_006254809.2 | XP_006254871.1 | 10 - 11 | 1560 | 103 | ||
XM_006254811.2 | XP_006254873.1 | 10 - 11 | 1689 | 103 | ||
GenBank mRNA | AB199794.1 | - | 10 - 11 | 1594 | 103 | |
BC168236.1 | - | 1636 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
