Assay Details
Gene Symbol: Rnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008299.2 | NP_001008300.1 | 8 - 9 | 1306 | 71 | |
XM_006254894.1 | XP_006254956.1 | 8 - 9 | 1467 | 71 | ||
XM_006254895.2 | XP_006254957.1 | 8 - 9 | 2741 | 71 | ||
XM_006254896.2 | XP_006254958.1 | 7 - 8 | 1163 | 71 | ||
GenBank mRNA | BC085858.1 | - | 8 - 9 | 1306 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
