Assay Details
Gene Symbol: Fech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108434.1 | NP_001101904.1 | 8 - 9 | 1003 | 76 | |
XM_006254850.2 | XP_006254912.1 | 8 - 9 | 3257 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
