Assay Details
Gene Symbol: Otub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106332.1 | NP_001099802.1 | 1 - 2 | 84 | 94 | |
GenBank mRNA | BC166771.1 | - | 1 - 2 | 67 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
