Assay Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231208.2 | XP_006231270.1 | 4 - 5 | 578 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
