Assay Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013152.1 | NP_001013170.1 | 2 - 3 | 378 | 89 | |
XM_006231208.2 | XP_006231270.1 | 6 - 7 | 824 | 89 | ||
GenBank mRNA | AY310155.1 | - | 814 | 89 | ||
AY325146.1 | - | 814 | 89 | |||
BC091413.1 | - | 2 - 3 | 378 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
