Assay Details
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053445.2 | NP_445897.2 | 11 - 12 | 1364 | 87 | |
XM_006231075.2 | XP_006231137.1 | 11 - 12 | 1511 | 87 | ||
GenBank mRNA | AB052085.1 | - | 11 - 12 | 1364 | 87 | |
AF320509.1 | - | 11 - 12 | 1393 | 87 | ||
FQ218734.1 | - | 10 - 11 | 1297 | 87 | ||
HQ909026.1 | - | 11 - 12 | 1288 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
