Assay Details
Gene Symbol: Nmrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024292.1 | NP_001019463.1 | 2 - 3 | 116 | 82 | |
XM_008760290.1 | XP_008758512.1 | 2 - 3 | 248 | 82 | ||
GenBank mRNA | BC079144.1 | - | 2 - 3 | 116 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079144.1, NM_001024292.1, XM_008760290.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
