Assay Details
Gene Symbol: Nmrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024292.1 | NP_001019463.1 | 4 - 5 | 256 | 106 | |
XM_008760290.1 | XP_008758512.1 | 4 - 5 | 388 | 106 | ||
GenBank mRNA | BC079144.1 | - | 4 - 5 | 256 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
