Assay Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271089.2 | NP_001258018.1 | 3 - 4 | 781 | 65 | |
NM_019283.3 | NP_062156.2 | 4 - 5 | 906 | 65 | ||
GenBank mRNA | AB015433.1 | - | 4 - 5 | 872 | 65 | |
BC061989.1 | - | 3 - 4 | 764 | 65 | ||
CV109741.1 | - | 2 - 3 | 123 | 65 | ||
U59324.1 | - | 3 - 4 | 695 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
