Assay Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271089.2 | NP_001258018.1 | 7 - 8 | 1231 | 86 | |
NM_019283.3 | NP_062156.2 | 8 - 9 | 1356 | 86 | ||
GenBank mRNA | AB015433.1 | - | 8 - 9 | 1322 | 86 | |
BC061989.1 | - | 7 - 8 | 1214 | 86 | ||
CV109741.1 | - | 6 - 7 | 573 | 86 | ||
U59324.1 | - | 7 - 8 | 1145 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
