Assay Details
Gene Symbol: Frmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008348.1 | NP_001008349.1 | 3 - 4 | 411 | 68 | |
XM_006230804.2 | XP_006230866.1 | 3 - 4 | 435 | 68 | ||
GenBank mRNA | BC085896.1 | - | 3 - 4 | 411 | 68 | |
FQ211651.1 | - | 3 - 4 | 450 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
