Assay Details
Gene Symbol: Frmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008348.1 | NP_001008349.1 | 9 - 10 | 1231 | 80 | |
XM_006230804.2 | XP_006230866.1 | 9 - 10 | 1255 | 80 | ||
GenBank mRNA | BC085896.1 | - | 9 - 10 | 1231 | 80 | |
FQ211651.1 | - | 9 - 10 | 1270 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
