Assay Details
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047854.2 | NP_001041319.2 | 3 - 4 | 331 | 79 | |
XM_006230548.2 | XP_006230610.1 | 3 - 4 | 405 | 79 | ||
XM_006230549.2 | XP_006230611.1 | 4 - 5 | 431 | 79 | ||
GenBank mRNA | AY325242.1 | - | 3 - 4 | 303 | 79 | |
BC127546.1 | - | 3 - 4 | 331 | 79 | ||
FQ213486.1 | - | 3 - 4 | 353 | 79 | ||
FQ228688.1 | - | 4 - 5 | 394 | 79 |
Gene Symbol: LOC100911881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230603.2 | XP_006230665.1 | 405 | 79 | ||
XM_006230604.2 | XP_006230666.1 | 431 | 79 | |||
XM_008760034.1 | XP_008758256.1 | 405 | 79 | |||
XM_008760035.1 | XP_008758257.1 | 427 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
