Assay Details
Gene Symbol: Gna14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013151.1 | NP_001013169.1 | 4 - 5 | 1078 | 83 | |
XM_008760288.1 | XP_008758510.1 | 5 - 6 | 1715 | 83 | ||
GenBank mRNA | BC090316.1 | - | 4 - 5 | 1078 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
