Assay Details
Gene Symbol: mrpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006973.1 | NP_001006974.1 | 2 - 3 | 261 | 148 | |
XM_006230739.2 | XP_006230801.1 | 2 - 3 | 305 | 148 | ||
GenBank mRNA | BC083740.1 | - | 2 - 3 | 261 | 148 | |
FQ225172.1 | - | 2 - 3 | 286 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
