Assay Details
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012197.2 | NP_001012197.2 | 11 - 12 | 1492 | 61 | |
GenBank mRNA | BC081917.1 | - | 11 - 12 | 1492 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
