Assay Details
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012197.2 | NP_001012197.2 | 13 - 14 | 2018 | 82 | |
GenBank mRNA | EV771986.1 | - | 1 - 2 | 293 | 82 | |
FQ229309.1 | - | 2 - 3 | 432 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
