Assay Details
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108101.1 | NP_001101571.1 | 10 - 11 | 1332 | 71 | |
GenBank mRNA | BC166731.1 | - | 10 - 11 | 1407 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
