Assay Details
Gene Symbol: Etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008344.1 | NP_001008345.1 | 6 - 7 | 832 | 113 | |
GenBank mRNA | BC085902.1 | - | 6 - 7 | 832 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
