Assay Details
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053398.1 | NP_445850.1 | 3 - 4 | 525 | 88 | |
GenBank mRNA | AF184920.1 | - | 1 - 2 | 65 | 88 | |
BC079378.1 | - | 3 - 4 | 525 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
