Assay Details
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053398.1 | NP_445850.1 | 6 - 7 | 1071 | 71 | |
GenBank mRNA | AF184920.1 | - | 4 - 5 | 611 | 71 | |
BC079378.1 | - | 6 - 7 | 1071 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
