Assay Details
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014111.1 | NP_001014133.1 | 3 - 4 | 309 | 103 | |
XM_006256835.2 | XP_006256897.2 | 4 - 5 | 695 | 103 | ||
XM_006256836.2 | XP_006256898.2 | 4 - 5 | 698 | 103 | ||
XM_006256839.2 | XP_006256901.2 | 4 - 5 | 695 | 103 | ||
XM_008773169.1 | XP_008771391.1 | 4 - 5 | 698 | 103 | ||
GenBank mRNA | BC086548.1 | - | 3 - 4 | 309 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
