Assay Details
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014111.1 | NP_001014133.1 | 7 - 8 | 845 | 140 | |
XM_006256835.2 | XP_006256897.2 | 8 - 9 | 1231 | 140 | ||
XM_006256836.2 | XP_006256898.2 | 8 - 9 | 1234 | 140 | ||
XM_006256839.2 | XP_006256901.2 | 8 - 9 | 1231 | 140 | ||
XM_008773169.1 | XP_008771391.1 | 8 - 9 | 1234 | 140 | ||
GenBank mRNA | BC086548.1 | - | 7 - 8 | 845 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
